Skip to Content
Merck
All Photos(1)

Key Documents

435R-1

Sigma-Aldrich

MCM3 (EP202) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP202, monoclonal

description

(For In Vitro Diagnostic Use in Select Regions (See Chart))

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (435R-14)
vial of 0.5 mL concentrate (435R-15)
bottle of 1.0 mL predilute (435R-17)
vial of 1.0 mL concentrate (435R-16)
bottle of 7.0 mL predilute (435R-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... MCM3(4172)

General description

MCM3, minichromosome maintenance protein 3, belongs to the MCM family of highly conserved group of DNA-binding proteins known to have a role in the initiation and regulation of DNA replication during the cell cycle.1-7 MCM3 expression is up-regulated in proliferating cells and absent in terminally differentiated cells. Like Ki-67, MCM3 marks proliferating cells. In a study performed by Endl E., et al., Ki-67 and MCM3 double staining was performed on human tonsil showing that MCM3 stains proliferating cells like Ki-67, and in addition marks cells present in the intermediate layer of epithelium (which are typically negative for Ki-67). Both Ki-67 and MCM3 are not detectable in differentiated cells.6 However, unlike Ki-67, MCM3 protein down-regulation is delayed. This suggests that Ki-67 may be expressed during a shorter interval of the cell cycle than MCM3. Anti-MCM3 may be a more reliable proliferation marker than anti-Ki-67.1

Quality


IVD

IVD

IVD

RUO

Linkage

MCM3 Positive Control Slides, Product No. 435S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Musahl et al.
Experimental cell research, 241(1), 260-264 (1998-06-20)
Mcm proteins are abundant nuclear proteins involved in the regulation of genome replication. Previous experiments had shown that levels of Mcm-specific mRNAs increase at the G1/S phase transition of the cell cycle, but that the amounts of Mcm proteins do
Mitali Das et al.
Molecular biology international, 2014, 574850-574850 (2014-11-12)
As a crucial component of DNA replication licensing system, minichromosome maintenance (MCM) 2-7 complex acts as the eukaryotic DNA replicative helicase. The six related MCM proteins form a heterohexamer and bind with ORC, CDC6, and Cdt1 to form the prereplication
Thilo Gambichler et al.
Journal of the American Academy of Dermatology, 60(5), 808-813 (2009-04-25)
Markers identifying premalignant and malignant melanocytic skin lesions are needed. We aimed to investigate the protein expression of minichromosome maintenance (MCM) proteins in melanocytic skin lesions with different malignant potential. Paraffin-embedded sections of benign melanocytic nevi (BN, n = 37)
The hexameric eukaryotic MCM helicase: building symmetry from nonidentical parts.
B K Tye et al.
The Journal of biological chemistry, 275(45), 34833-34836 (2000-09-12)
E Endl et al.
The Journal of pathology, 195(4), 457-462 (2001-12-18)
The mini-chromosome maintenance proteins (MCM), which are involved in the control of DNA replication, and the cyclin-dependent kinase inhibitors, such as p27/KIP1, represent two groups of proteins that are currently under investigation as diagnostic tumour markers. The expression of p27

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service