Skip to Content
Merck
All Photos(1)

Key Documents

164114K

Sigma-Aldrich

Laminin Mimetic animal-free ECM peptides

MAPTRIX-L-RYVVLPR, solution, 1.0 mg/mL protein, suitable for cell culture

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€473.00

€473.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€473.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.75

€473.00


Estimated to ship on20 May 2025


Request a Bulk Order

Product Name

MAPTRIX-L-RYVVLPR, Laminin Mimetic, aqueous solution

sterility

Filtered sterilized solution

form

aqueous solution

mol wt

~23.5 kDa

concentration

1.0 mg/mL protein

technique(s)

cell culture | mammalian: suitable

Binding Specificity

Receptor: Heparin
Domain: β1 chain - F9
Binding Motif: RYVVLPR

shipped in

ambient

storage temp.

2-8°C

Application

MAPTrix Technology provides a true extracellular microenvironment (ECM) by presenting combinatorial peptide motifs. The ECM, defined by biochemical cues and physical cues, is a deciding factor in a wide range of cellular processes including cell adhesion, proliferation, differentiation, and expression of phenotype-specific functions.Currently existing technology offers simple and merely adequate environments that facilitate simple cell processes such as cellular adhesion.[1][2] The simple presentation of cell adhesion motifs is not optimal for controlling more integrated processes. Crosstalk among signaling pathways act synergistically to enhance cellular responses such as cell adhesion or proliferation.[3] A study showed that a combination of extracellular matrix derived peptides presented on a surface may enhance cell adhesion strength and focal adhesion assembly. The combinatorial presentation of ECM peptides on cell growth surfaces may also promote elevated proliferation rates of primary or stem cells.[4][5][6]

Features and Benefits

  • Biochemically-defined, animal-free cell culture surfaces designed to enhance cell performance
  • MAPTrix produces a uniform ECM surface that provides a highly controlled 2D extracellular microenvironment for cell cultures and related applications
  • Adhesion properties of the mussel adhesive protein makes the coating reproducible & reliable

Legal Information

MAPTrix is a trademark of Kollodis Biosciences

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samuel Schmidt et al.
Cell and tissue research, 339(1), 83-92 (2009-11-19)
Adhesion and migration are integrated cell functions that build, maintain and remodel the multicellular organism. In migrating cells, integrins are the main transmembrane receptors that provide dynamic interactions between extracellular ligands and actin cytoskeleton and signalling machineries. In parallel to
Seung Tae Lee et al.
Biomaterials, 31(6), 1219-1226 (2009-11-21)
We present development and use of a 3D synthetic extracellular matrix (ECM) analog with integrin-specific adhesion ligands to characterize the microenvironmental influences in embryonic stem cell (ESC) self-renewal. Transcriptional analysis of 24 integrin subunits followed by confirmation at the translational
Stephanie J Ellis et al.
Cell and tissue research, 339(1), 121-130 (2009-07-10)
The regulation of stem cell behavior and maintenance typically involves the integration of both intrinsic and extrinsic cues. One such external cue, integrin-mediated cell adhesion to the extracellular matrix, plays an important part in regulating stem cell function and maintenance.
S J Fashena et al.
Nature cell biology, 2(12), E225-E229 (2001-01-09)
Over the past twenty years, intensive research has enabled us to identify components of specific signalling pathways downstream of an array of adhesion and growth-factor receptors. The first Gordon Research Conference on 'Signalling by Adhesion Receptors', which took place in
Catherine D Reyes et al.
Journal of cellular physiology, 217(2), 450-458 (2008-07-10)
Cell adhesion to extracellular matrix (ECM) components through cell-surface integrin receptors is essential to the formation, maintenance and repair of numerous tissues, and therefore represents a central theme in the design of bioactive materials that successfully interface with the body.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service