Skip to Content
Merck
All Photos(1)

Key Documents

15931

Sigma-Aldrich

Hexaethyleneglycol monododecyl ether 1 mM solution

Synonym(s):

Hexaethyleneglycol monododecyl ether solution, Detergent Screening Solution 18/Kit-No 66317

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

description

non-ionic

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCOCCOCCOCCOCCOCCOCCO


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service