Skip to Content
Merck
All Photos(1)

Key Documents

137275

SAFC

Glycine hydrochloride

EMPROVE® EVOLVE

Pharma Manufacturing

Synonym(s):

Glycine hydrochloride, Aminoacetic acid, Aminoethanoic acid, Glycocoll

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€185.00
1 KG
€540.00
10 KG
€4,884.00
25 KG
€10,616.00
50 KG
€20,007.00

€185.00


Estimated to ship on18 April 2025



Select a Size

Change View
100 G
€185.00
1 KG
€540.00
10 KG
€4,884.00
25 KG
€10,616.00
50 KG
€20,007.00

About This Item

Linear Formula:
H2NCH2CO2H · HCl
CAS Number:
Molecular Weight:
111.53
MDL number:
UNSPSC Code:
12352209

€185.00


Estimated to ship on18 April 2025


biological source

synthetic

Quality Level

product line

EMPROVE® EVOLVE

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, microbial, heavy metals, arsenic and trace impurities; tested

mp

176-180 °C (lit.)

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity; tested

storage temp.

2-30°C

SMILES string

Cl.NCC(O)=O

InChI

1S/C2H5NO2.ClH/c3-1-2(4)5;/h1,3H2,(H,4,5);1H

InChI key

IVLXQGJVBGMLRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service