Skip to Content
Merck
All Photos(1)

Key Documents

01824

Sigma-Aldrich

Acylase I, immobilized on Eupergit® C from Aspergillus sp.

≥50 U/g moist material

Synonym(s):

Aminoacylase, immobilized, Plexazym® AC

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

form

beads

specific activity

≥50 U/g moist material

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

the immobilized acylase catalyzes the hydrolysis of N-acetyl-DL-amino acid to L-amino acid, the D-form is not attacked

Unit Definition

1 U corresponds to the amount of enzyme which hydrolyzes 1 μmol N-acetyl-L-methionine per minute at pH 8.0 and 25°C

Reconstitution

Standard procedure: a 10-20% substrate solution, pH 6-8, with an addition of CoCl2 (10-4 moles) at 33°C was used. Prior to use the polymer was washed with water (50 times bed volumes); when used in a fixed bed reactor, a velocity of flow of 3 bed volumes/h leads to a hydrolysis degree of 80%

Analysis Note

moist pearls (dried substance ~30%, pearl diameter 50-100 μm), covalent fixation of the acylase

Other Notes

The immobilized acylase is used for the convenient resolution of amino acids via the selective deacetylation of N-acetyl-L-amino acids in DL-racemates[1][2][3][4]

Legal Information

Eupergit is a registered trademark of Röhm GmbH & Co. KG
Plexazym is a registered trademark of Röhm GmbH & Co. KG

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Tramper
Solid Phase Biochemistry, 393-393 (1983)
Preparation and properties of enzymes immobilized by copolymerization.
D Jaworek et al.
Methods in enzymology, 44, 195-120 (1976-01-01)
Optical resolution of racemic amino acids by aminoacylase.
T Sato et al.
Bioprocess technology, 16, 3-14 (1993-01-01)
W. Kuhlmann et al.
Chemie Ingenieur Technik, 52, 607-607 (1980)
Keya Cai et al.
Sheng wu gong cheng xue bao = Chinese journal of biotechnology, 24(2), 285-290 (2008-05-10)
Three model silkworms, highly resistant strain, highly susceptible strain and their near isogenic line were established by hybridization and backcross. The resistance of silkworm (Bombyx mori L.) to BmNPV was studied at proteomic level using two-dimensional gel electrophoresis and MALDI

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service