Skip to Content
Merck
All Photos(1)

Key Documents

00409

Sigma-Aldrich

Iris 7-WS carboxylic acid

Synonym(s):

2-{2-{2-[4-(4-Carboxybutyrylamino)phenoxy]-3-{2-[1-(hex-5-ynyl)-1,3-dihydro-3,3-dimethyl-5-sulfoindol-2-ylidene]ethylidene}cyclohex-1-enyl}vinyl}-1-(hex-5-ynyl)-3,3-dimethyl-3H-indolium-5-sulfonate monopotassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H58KN3O10S2
CAS Number:
Molecular Weight:
1000.27
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

fluorescence

λex 777 nm; λem 796 nm in 0.1 M phosphate pH 7.0

storage temp.

−20°C

SMILES string

[K+].CC1(C)/C(=C\C=C2/CCCC(\C=C\C3=[N+](CCCCC#C)c4ccc(cc4C3(C)C)S([O-])(=O)=O)=C2Oc5ccc(NC(=O)CCCC(O)=O)cc5)N(CCCCC#C)c6ccc(cc16)S([O-])(=O)=O

InChI

1S/C53H59N3O10S2.K/c1-7-9-11-13-33-55-45-29-27-41(67(60,61)62)35-43(45)52(3,4)47(55)31-21-37-17-15-18-38(51(37)66-40-25-23-39(24-26-40)54-49(57)19-16-20-50(58)59)22-32-48-53(5,6)44-36-42(68(63,64)65)28-30-46(44)56(48)34-14-12-10-8-2;/h1-2,21-32,35-36H,9-20,33-34H2,3-6H3,(H3-,54,57,58,59,60,61,62,63,64,65);/q;+1/p-1

InChI key

YQXFZXZEZDXDBU-UHFFFAOYSA-M

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Principle

The dye can be activated at the carboxy group for coupling purposes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiara A Bertolino et al.
Journal of fluorescence, 16(2), 221-225 (2006-05-03)
A series of novel functionalized, water-soluble, pH-unsensitive, highly photostable heptamethine cyanine dyes (HCDs) has been synthesized. The aim of the synthesis was to obtain novel effective probes for fluorescence detection in the near infrared. Synthesis and characterization of a special

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service