VT000834
Enterobacter cloacae WDCM 00083 Vitroids
130 - 300 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Certified reference material, Microbiological standards, Enterobacter WDCM 00083 Vitroids
About This Item
Recommended Products
grade
certified reference material
Quality Level
Agency
ISO 17034
ISO/IEC 17025
form
disc (hydrosoluble)
solid
shelf life
limited shelf life, expiry date on the label
packaging
package of 10 discs
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
application(s)
environmental
food and beverages
microbiology
storage temp.
−20°C
suitability
Enterobacter cloacae
Enterobacter spp.
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The technology allows long-term conservation and renders the organisms resistant to temperature changes. They are intended to be used as daily internal quality control organisms.
These microorganisms are certified according to ISO/IEC 17025 and produced under reproducible conditions compliant with ISO 17034 using authenticated strains from CECT®. These ready-to-use microbiological controls minimize the need for maintaining control strains in the test laboratory and guarantee the use of an authenticated control culture for every quality control test.
Vitroids are derived from a traceable culture obtained freeze-dried from CECT®.
Application
Features and Benefits
- Ready-to-use at convenient CFU ranges
- No dilution required
- Quick rehydration in 10 minutes
- 2-3 years of shelf life in a standard -20 °C freezer
- Complete with a comprehensive certificate of analysis
- No maintenance of stock cultures
- No recovery time or pre-enrichment step is necessary
- Robust, efficient, and reliable
- Easy-to-use
- Cost-effective
Other Notes
Other cell culture collections:
- DSM 30054
- ATCC® 13047
- NCTC 10005
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Explore certified microbiological reference materials: VITROIDS™, LENTICULE® discs by Millipore®. Ideal for food, beverage, cannabis testing. Order today.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service