Skip to Content
Merck
All Photos(1)

Key Documents

TSS5

Supelco

Total Suspended Solids 5mg/L

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

100
300

CofA

current certificate can be downloaded

application(s)

environmental

format

single component solution

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Total Suspended Solids 5mg/L for R&D and Analytical use only.

Features and Benefits

  • Product is of Certified Reference Material grade
  • Packed in High Density Polyethylene (HDPE) bottle with 500ml of solution
  • A certificate of analysis (CoA) is included with the product
  • Traceable to recognized national standards, and units of measurement recognized at the National Institute of Standards and Technology (NIST) and international system of units (SI)
  • Gravimetrically performed dilutions
  • Balance used to prepare the standard is calibrated to ISO 17025 requirements
  • Stable solution with long minimum shelf life

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service