Skip to Content
Merck
All Photos(1)

Documents

T8258

Sigma-Aldrich

Trifluoroacetic anhydride

liquid

Synonym(s):

TFAA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3CO)2O
CAS Number:
Molecular Weight:
210.03
Beilstein:
746197
EC Number:
MDL number:
UNSPSC Code:
12352100

vapor pressure

6.28 psi ( 20 °C)

form

liquid

color

clear colorless

refractive index

n20/D 1.3 (lit.)

bp

39.5-40 °C (lit.)

mp

−65 °C (lit.)

density

1.511 g/mL at 20 °C (lit.)

SMILES string

FC(F)(F)C(=O)OC(=O)C(F)(F)F

InChI

1S/C4F6O3/c5-3(6,7)1(11)13-2(12)4(8,9)10

InChI key

QAEDZJGFFMLHHQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angelos Halaris et al.
Journal of psychiatric research, 66-67, 118-126 (2015-05-27)
A pro-inflammatory state and a dysregulation in the tryptophan/kynurenine pathway have been documented in depression. This study examined whether treatment with the SSRI, escitalopram (ESC), could suppress inflammation and favorably shift metabolites of the kynurenine pathway in patients with major
Hidenori Kinoshita et al.
Journal of the American Chemical Society, 132(18), 6412-6418 (2010-04-17)
Reaction screening of nucleophilic reaction partners for addition to N-diphenylphosphinylimines employing lanthanum(III) triflate as a catalyst and trifluoroacetic anhydride (TFAA) as an activator is reported. A number of tandem processes leading to novel chemotypes including aza-Prins/intramolecular Friedel-Crafts annulations have been
Philippe Nuhant et al.
The Journal of organic chemistry, 74(24), 9413-9421 (2009-11-21)
Aminopentadienals resulting from the condensation of tryptamine or homoveratrylamine with glutaconaldehydes were treated with trifluoroacetic anhydride, allowing the formation of tetrahydro-beta-carbolines and tetrahydroisoquinolines bearing an enal function. In this N-acyl Pictet-Spengler reaction the electrophilicity of the aminopentadienals was dramatically increased
Lok Ranjan Bhatt et al.
Carbohydrate research, 346(5), 691-694 (2011-03-01)
Acylation of chitin with butyric acid was performed in the presence of trifluoroacetic anhydride/phosphoric acid mediated system. The products were characterized by (1)H NMR and FT-IR spectroscopy and their solubility was tested in different organic solvents. Inclusion of butyric acid
Bjorn J A Berendsen et al.
Analytica chimica acta, 585(1), 126-133 (2007-03-28)
Derivatisation of the avermectines ivermectin (IVM), doramectin (DOR), abamectin (ABA) and eprinomectin (EPR), and the milbemycin moxidectin (MOX) to fluorescent derivatives is commonly used for quantitative analysis at relevant levels using high performance liquid chromatography (HPLC) with fluorescence detection. Problems

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service