Skip to Content
Merck
All Photos(1)

Key Documents

SMB00950

Sigma-Aldrich

O-Desmethyl gefitinib

≥95%

Synonym(s):

4-[(3-Chloro-4-fluorophenyl)amino]-6-[3-(4-morpholinyl)propoxy]-7-quinazolinol, O-Desmethylgefitinib

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22ClFN4O3
CAS Number:
Molecular Weight:
432.88
UNSPSC Code:
12352205
NACRES:
NA.77

biological source

synthetic

Quality Level

grade

research grade

Assay

≥95%

form

solid

technique(s)

HPLC: suitable

color

light yellow to yellow

storage temp.

−20°C

InChI

1S/C21H22ClFN4O3/c22-16-10-14(2-3-17(16)23)26-21-15-11-20(19(28)12-18(15)24-13-25-21)30-7-1-4-27-5-8-29-9-6-27/h2-3,10-13,28H,1,4-9H2,(H,24,25,26)

InChI key

IFMMYZUUCFPEHR-UHFFFAOYSA-N

General description

O-Desmethyl Gefitinib, a key metabolite of the anticancer drug gefitinib, belongs to the quinazoline family and is present in human plasma. Its formation is facilitated by the cytochrome P450 isoform CYP2D6. This compound exhibits significant activity as an inhibitor of tyrosine residue phosphorylation in EGFR, with an IC50 of 36 nM in subcellular assays. While its effectiveness is comparable to that of gefitinib, O-Desmethyl Gefitinib shows slightly reduced activity in whole-cell assays, with an IC50 of 760, in contrast to gefitinib′s 49 nM. Studies conducted in LoVo tumor mouse xenograft models have revealed that the tumor concentration of O-Desmethyl Gefitinib is notably lower than that of gefitinib, without causing a substantial reduction in tumor growth. Remarkably, increased plasma concentrations of O-Desmethyl Gefitinib in individuals homozygous for CYP2D6 do not lead to an increase in adverse effects. O-Desmethyl Gefitinib holds promise for biochemical and biomedical research, providing potential insights into drug development and cancer treatment.

Application

O-Desmethyl Gefitinib finds application in metabolomics and drug discovery research, particularly in the investigation of cancer and tumor management.

Biochem/physiol Actions

O-Desmethyl gefitinib is generated by the removal of a methyl group from gefitinib during the body′s metabolic processing of this compound within the body.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with a wide variety of chromatographic and spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.
This product is intended for research purposes only, and it is not meant for human consumption.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minimal contribution of desmethyl-gefitinib, the major human plasma metabolite of gefitinib, to epidermal growth factor receptor (EGFR)-mediated tumour growth inhibition
McKillop et al.
Xenobiotica, 36, 29-39 (2006)
Hiroyuki Kobayashi et al.
Medical oncology (Northwood, London, England), 33(6), 57-57 (2016-05-08)
We investigated the effects of polymorphisms in CYP2D6, ABCB1, and ABCG2 and the side effects induced by gefitinib on the pharmacokinetics of O-desmethyl gefitinib, the active metabolite of gefitinib. On day 14 after beginning therapy with gefitinib, plasma concentrations of
Metabolic disposition of gefitinib, an epidermal growth factor receptor tyrosine kinase inhibitor, in rat, dog and man
McKillop et al.
Xenobiotica, 34, 917-934 (2004)
Ping Fang et al.
Drug design, development and therapy, 11, 1283-1290 (2017-05-04)
Cytochrome P450 2D6 (CYP2D6), a member of the CYP450 enzyme super family, is a polymorphic enzyme that metabolizes ~25% of therapeutic drugs. CYP2D6 exhibits significant genetic polymorphisms which might cause adverse effects and therapeutic failures of some drugs. The purpose
D McKillop et al.
Xenobiotica; the fate of foreign compounds in biological systems, 34(10), 901-915 (2005-03-15)
The pharmacokinetics of gefitinib and its metabolites in rat and dog were investigated in preclinical studies conducted to support the safety evaluation and clinical development of gefitinib, the first EGFR tyrosine kinase inhibitor approved for the treatment of non-small-cell lung

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service