Skip to Content
Merck
All Photos(1)

Key Documents

R6006

Sigma-Aldrich

Remazol Brilliant Violet 5R

Synonym(s):

Reactive Violet 5

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Colour Index Number:
18097
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

SMILES string

[Na+].[Na+].[Na+].CC(=O)Nc1cc(cc2cc(c(\N=N\c3cc(ccc3O)S(=O)(=O)CCOS([O-])(=O)=O)c(O)c12)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C20H19N3O15S4.3Na/c1-10(24)21-15-9-13(40(29,30)31)6-11-7-17(41(32,33)34)19(20(26)18(11)15)23-22-14-8-12(2-3-16(14)25)39(27,28)5-4-38-42(35,36)37;;;/h2-3,6-9,25-26H,4-5H2,1H3,(H,21,24)(H,29,30,31)(H,32,33,34)(H,35,36,37);;;/q;3*+1/p-3/b23-22+;;;

InChI key

UBTDYBUJBRAYEZ-DQUNTQLESA-K

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chirayu Desai et al.
Biodegradation, 20(6), 813-826 (2009-06-12)
Bacterial consortium-AIE2 with a capability of contemporaneous Cr(VI) reduction and azo dye RV5 decolourization was developed from industrial wastewaters by enrichment culture technique. The 16S rRNA gene based molecular analyses revealed that the consortium bacterial community structure consisted of four
K Vijayaraghavan et al.
Journal of hazardous materials, 167(1-3), 790-796 (2009-02-24)
A complex Remazol dye effluent, comprised of four reactive dyes and auxiliary chemicals, was decolorized using SPS-200 (sawdust-based) and SPC-100 (coal-based) activated carbons. A detailed characterization revealed that the pore diameter of the activated carbon played an important role in
Adriana P Vieira et al.
Journal of hazardous materials, 166(2-3), 1272-1278 (2009-01-20)
Extracted babassu coconut (Orbignya speciosa) mesocarp (BCM) was applied as a biosorbent for aqueous Blue Remazol R160 (BR 160), Rubi S2G (R S2G), Red Remazol 5R (RR 5), Violet Remazol 5R (VR 5) and Indanthrene Olive Green (IOG) dye solutions.
N D Lourenço et al.
Journal of biotechnology, 89(2-3), 163-174 (2001-08-14)
The combination of anaerobic and aerobic periods in the operation cycle of a Sequencing Batch Reactor (SBR) was chosen to study biological color removal from simulated textile effluents containing reactive, sulfonated, monoazo and diazo dyes, respectively, Remazol Brilliant Violet 5R
Kevser Cirik et al.
Bioprocess and biosystems engineering, 36(1), 69-79 (2012-07-28)
The aim of the study was to investigate the effect of nitrate on anaerobic color removal efficiencies. For this aim, anaerobic-aerobic sequencing batch reactor (SBR) fed with a simulated textile effluent including Remazol Brilliant Violet 5R azo dye was operated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service