Skip to Content
Merck
All Photos(1)

Key Documents

QC1456

Supelco

Trihalomethanes - WS

certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

1.2 ML
€82.00

€82.00


Available to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
1.2 ML
€82.00

About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

€82.00


Available to ship on07 April 2025Details


Request a Bulk Order

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

packaging

ampule of 1.2 mL

manufacturer/tradename

RTC QCO-002

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental
food and beverages

format

matrix material

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

    Bromodichloromethane
    Bromoform
    Chloroform
    Dibromochloromethane

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

WGK

WGK 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service