PHR8934
Melatonin-formaldehyde condensation product
Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
N,N′-[Methylenebis[(5-methoxy-1H-indole-1,3-diyl)-2,1-ethanediyl]]bis-acetamide, N,N′-[Methylenebis[(5-methoxy-1H-indole-1,3-diyl)-2,1-ethanediyl]]bis-acetamide
About This Item
grade
certified reference material
pharmaceutical secondary standard
Quality Level
API family
melatonin
form
solid
packaging
pkg of 50 mg
application(s)
pharmaceutical small molecule
Related Categories
General description
Application
Analysis Note
Other Notes
Recommended products
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service