Skip to Content
Merck
All Photos(1)

Documents

PHR1704

Supelco

Nafcillin Sodium

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Nafcillin sodium salt monohydrate, 6-(2-Ethoxy-1-naphthamido)penicillin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H21N2O5SNa · H2O
CAS Number:
Molecular Weight:
454.47
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1450007

API family

nafcillin

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

-10 to -25°C

SMILES string

O.[Na+].CCOc1ccc2ccccc2c1C(=O)N[C@H]3C4SC(C)(C)[C@@H](N4C3=O)C([O-])=O

InChI

1S/C21H22N2O5S.Na.H2O/c1-4-28-13-10-9-11-7-5-6-8-12(11)14(13)17(24)22-15-18(25)23-16(20(26)27)21(2,3)29-19(15)23;;/h5-10,15-16,19H,4H2,1-3H3,(H,22,24)(H,26,27);;1H2/q;+1;/p-1/t15-,16+,19-;;/m1../s1

InChI key

OCXSDHJRMYFTMA-KMFBOIRUSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Nafcillin Sodium is a narrow-spectrum β-lactamase resistant antibiotic drug.

Application

Nafcillin Sodium may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

related product

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sensitive ion chromatographic determination of citrate and formate in pharmaceuticals
Kaleemullah T, et al.
Rasayan Journal of Chemistry, 4(4), 844-852 (2011)
Nafcillin Sodium
USP42-NF37
United States Pharmacopeia/National Formulary, 3029-3029 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service