Skip to Content
Merck
All Photos(1)

Documents

P4419

Supelco

Pridinol methanesulfonate salt

analytical standard

Synonym(s):

α,α-Diphenyl-1-piperidinepropanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H25NO · CH4O3S
CAS Number:
Molecular Weight:
391.52
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

CS(O)(=O)=O.OC(CCN1CCCCC1)(c2ccccc2)c3ccccc3

InChI

1S/C20H25NO.CH4O3S/c22-20(18-10-4-1-5-11-18,19-12-6-2-7-13-19)14-17-21-15-8-3-9-16-21;1-5(2,3)4/h1-2,4-7,10-13,22H,3,8-9,14-17H2;1H3,(H,2,3,4)

InChI key

VNJHUUNVDMYCRH-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Antiparkinsonian

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Comparison of systemic and local therapy in rheumatic myalagia and arthralgia].
G Fraschini
La Clinica terapeutica, 99(3), 305-312 (1981-11-15)
Peter Svensson et al.
European journal of pain (London, England), 7(5), 449-456 (2003-08-26)
A randomised, double-blind, placebo-controlled three-way cross-over study was performed to investigate the effect of two muscle relaxants (tolperisone hydrochloride and pridinol mesilate) on experimental jaw-muscle pain and jaw-stretch reflexes. Fifteen healthy men participated in three randomised sessions separated by at
[Possibility of occurrence of psychoses after pridinol treatment].
Z Cichecki
Psychiatria polska, 14(1), 88-89 (1980-01-01)
Pridinolum mesylate and neuroleptic malignant syndrome.
L Giordani et al.
The American journal of psychiatry, 142(3), 389-390 (1985-03-01)
[Clinical study of a drug combination in the treatment of metarheumatic and metatraumatic disorders].
A Cogliandro
La Clinica terapeutica, 101(3), 267-274 (1982-05-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service