Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 100 g
manufacturer/tradename
NIST®
technique(s)
XRF: suitable
optical emission spectroscopy (OES): suitable
format
matrix material
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
This Standard Reference Material (SRM) is intended primarily for use in the evaluation of chemical and instrumental methods of analysis of nickel alloys and similar matrices. A unit of SRM 864 is a single bottle containing 50 g of chips. SRM 1244 is Nickel Alloy UNS N06600 in disk form. For more information, please refer to the COA and SDS.
SRM 864_cert
SRM 864_SDS
SRM 864_cert
SRM 864_SDS
Other Notes
Certified for the analytes listed below.
Aluminum (Al), Cobalt (Co), Chromium (Cr), Copper (Cu), Iron (Fe), Manganese (Mn), Molybdenum (Mo) , Nickel (Ni) , Vanadium (V), Boron (B), Magnesium (Mg), Lead (Pb), Thallium (Tl)
Reference/Informational value is provided for the following analytes. Carbon (C), Niobium (Nb), Phosphorus (P), Sulfur (S), Silicon (Si), Titanium (Ti), Arsenic (As), Tin (Sn), Zirconium (Zr), Silver (Ag), Bismuth (Bi), Calcium (Ca), Gallium (Ga), Nitrogen (N), Oxygen (O), Antimony (Sb), Selenium (Se), Tantalum (Ta), Tellurium (Te), Tungsten (W)
See certificate for values and more details at nist.gov/SRM.
Aluminum (Al), Cobalt (Co), Chromium (Cr), Copper (Cu), Iron (Fe), Manganese (Mn), Molybdenum (Mo) , Nickel (Ni) , Vanadium (V), Boron (B), Magnesium (Mg), Lead (Pb), Thallium (Tl)
Reference/Informational value is provided for the following analytes. Carbon (C), Niobium (Nb), Phosphorus (P), Sulfur (S), Silicon (Si), Titanium (Ti), Arsenic (As), Tin (Sn), Zirconium (Zr), Silver (Ag), Bismuth (Bi), Calcium (Ca), Gallium (Ga), Nitrogen (N), Oxygen (O), Antimony (Sb), Selenium (Se), Tantalum (Ta), Tellurium (Te), Tungsten (W)
See certificate for values and more details at nist.gov/SRM.
Legal Information
Inconel is a registered trademark of Special Metals Corporation
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
related product
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service