Skip to Content
Merck
All Photos(1)

Key Documents

NIST2630

Nitric oxide in nitrogen

NIST® SRM® 2630, nominal 1500 umol/mol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352303
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

packaging

pkg of each

manufacturer/tradename

NIST®

application(s)

environmental

format

mixture

SMILES string

N=O

InChI

1S/HNO/c1-2/h1H

InChI key

ODUCDPQEXGNKDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Other Notes

Certified for the analytes listed below.
Nitric oxide (NO)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service