Skip to Content
Merck
All Photos(1)

Documents

NIST1907

Nickel microhardness test block (Knoop)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of each

manufacturer/tradename

NIST®

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is intended primarily for use in calibrating Knoop-type microhardness testers. SRM 1907 consists of a 1.35 cm square test block of electrodeposited bright nickel (approx. 750 μm thick) on an AISI 1010 steel substrate. For more information, please refer to the COA and SDS.

SRM 1907_cert

SRM 1907_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Nickel (Ni)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service