Skip to Content
Merck
All Photos(1)

Key Documents

D4670

Sigma-Aldrich

Diethylene glycol

suitable for photographic applications

Synonym(s):

2,2′-Oxydiethanol, 2-Hydroxyethyl ether, Bis(2-hydroxyethyl) ether, Diglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2CH2)2O
CAS Number:
Molecular Weight:
106.12
Beilstein:
969209
EC Number:
MDL number:
UNSPSC Code:
12352100

vapor density

2.14 (vs air)

vapor pressure

0.01 mmHg ( 20 °C)

description

non-ionic

autoignition temp.

442 °F

expl. lim.

2-12.3 %

refractive index

n20/D 1.447 (lit.)

bp

245 °C (lit.)

mp

−10 °C (lit.)

density

1.118 g/mL at 25 °C (lit.)

suitability

suitable for photographic applications

SMILES string

OCCOCCO

InChI

1S/C4H10O3/c5-1-3-7-4-2-6/h5-6H,1-4H2

InChI key

MTHSVFCYNBDYFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ze-Jian Wang et al.
Journal of photochemistry and photobiology. B, Biology, 115, 16-24 (2012-07-17)
Photodynamic therapy (PDT) has recently been proposed as a possible indication in the conservative treatment of hereditary retinoblastoma. In order to create photosensitizers with enhanced targeting ability toward retinoblastoma cells, meso-tetraphenylporphyrins bearing one glycodendrimeric moiety have been synthesized. The binding
Parisa Sadatmousavi et al.
Acta biomaterialia, 8(9), 3241-3250 (2012-05-30)
Self-assembling peptide nanofibers have emerged as important nanobiomaterials, with such applications as delivery of therapeutic agents and vaccines, nanofabrication and biomineralization, tissue engineering and regenerative medicine. Recently a new class of self-assembling peptides has been introduced, which takes into consideration
Asuman C Kucuk et al.
Langmuir : the ACS journal of surfaces and colloids, 27(10), 6381-6388 (2011-04-16)
"Core-corona" type amphiphiles, which comprise double-decker-shaped POSSs (DDSQs) as the core and two or four di(ethylene glycol) (DEG) units as the coronae, have recently been reported to form a stable monolayer at the air-water interface. In this paper, another core-corona
Leo J Schep et al.
Clinical toxicology (Philadelphia, Pa.), 47(6), 525-535 (2009-07-10)
Diethylene glycol (DEG) is a clear, colorless, practically odorless, viscous, hygroscopic liquid with a sweetish taste. In addition to its use in a wide range of industrial products, it has also been involved in a number of prominent mass poisonings
Bichismita Sahu et al.
The Journal of organic chemistry, 76(14), 5614-5627 (2011-05-31)
Developed in the early 1990s, peptide nucleic acid (PNA) has emerged as a promising class of nucleic acid mimic because of its strong binding affinity and sequence selectivity toward DNA and RNA and resistance to enzymatic degradation by proteases and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service