Skip to Content
Merck
All Photos(1)

Documents

D3525

Supelco

Diosmin

analytical standard

Synonym(s):

3′,5,7-Trihydroxy-4′-methoxyflavone 7-rutinoside, Barosmin, Buchu resin, Diosmetin 7-O-rhamnosylglucoside, Diosmetin 7-rutinoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H32O15
CAS Number:
Molecular Weight:
608.54
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(cc1O)C2=CC(=O)c3c(O)cc(O[C@@H]4O[C@H](CO[C@@H]5O[C@@H](C)[C@H](O)[C@@H](O)[C@H]5O)[C@@H](O)[C@H](O)[C@H]4O)cc3O2

InChI

1S/C28H32O15/c1-10-21(32)23(34)25(36)27(40-10)39-9-19-22(33)24(35)26(37)28(43-19)41-12-6-14(30)20-15(31)8-17(42-18(20)7-12)11-3-4-16(38-2)13(29)5-11/h3-8,10,19,21-30,32-37H,9H2,1-2H3/t10-,19+,21-,22+,23+,24-,25+,26+,27+,28+/m0/s1

InChI key

GZSOSUNBTXMUFQ-YFAPSIMESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Citrus flavonoid. Modification of hesperidin. Studied for its chemopreventive properties, including anti-proliferative and anti-inflammatory.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jacqueline M Cohen et al.
Chest, 141(2), 308-320 (2012-02-09)
Postthrombotic syndrome (PTS) is a frequent, chronic complication of DVT. The effectiveness and safety of available treatments are unknown. The objective of this study was to systematically review the literature to assess whether pharmacologic and compression therapies are effective and
Mir Tahir et al.
Alcohol (Fayetteville, N.Y.), 47(2), 131-139 (2013-02-20)
The present investigation was designed to evaluate the efficacy of diosmin against ethanol-induced hepatotoxicity in rats by modulating various mechanisms including ethanol metabolizing enzymes, generation of free radicals, imbalance in oxidant-antioxidant status, oxidative damage to membrane lipids, activation of transcription
M Sinan Başoğlu et al.
International journal of pediatric otorhinolaryngology, 76(9), 1343-1346 (2012-07-06)
To evaluate the effectiveness of micronized flavonoid fraction in preventing cisplatin ototoxicity in a guinea pig model. This study was conducted on 23 guinea pigs in the Animal Laboratory of Izmir Atatürk Training and Research Hospital. Animals were divided into
Thangarasu Silambarasan et al.
European journal of pharmacology, 679(1-3), 81-89 (2012-01-24)
The present study was aimed to evaluate the antihypertensive effect of diosmin in deoxycorticosterone acetate (DOCA)-salt induced hypertension in male Wistar rats. Hypertension was induced in uninephrectomized rats by weekly twice subcutaneous injection of DOCA (25 mg/kg body weight) and
M Alcaraz et al.
Dento maxillo facial radiology, 40(5), 310-314 (2011-06-24)
The aim of this study was to evaluate the antioxidant substances present in the human diet with an antimutagenic protective capacity against genotoxic damage induced by exposure to X-rays in an attempt to reduce biological damage to as low a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service