CRM355
TPH - Sandy Loam 1
certified reference material, pkg of 100 g
Synonym(s):
Sand matrix CRM, TPH
About This Item
Recommended Products
grade
certified reference material
CofA
current certificate can be downloaded
feature
Certified TPH Concentration - 7040 mg/Kg
TPH Source: Gasoline/Diesel
packaging
pkg of 100 g
manufacturer/tradename
RTC CRM355-100
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
matrix
soil
pH
7.51
application(s)
environmental
format
matrix material
storage temp.
2-30°C
Related Categories
General description
Application
Other Notes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 1B
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service