BCR708
Dairy feed
BCR®, certified reference material
Synonym(s):
Feed for dairy cattle
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
85151701
Recommended Products
grade
certified reference material
Agency
BCR®
manufacturer/tradename
JRC
application(s)
agriculture
cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)
format
matrix material
storage temp.
−20°C
Analysis Note
For more information please see:
BCR708
BCR708
Other Notes
Certified for the analytes listed below. See certificate for values and more details
Crude ash, Crude fibre, Crude oils and fats, Phosphate, Crude protein
Trace Elements: Calcium (Ca), Copper (Cu), Magnesium (Mg)
Matrix Group: Animal Feed
Crude ash, Crude fibre, Crude oils and fats, Phosphate, Crude protein
Trace Elements: Calcium (Ca), Copper (Cu), Magnesium (Mg)
Matrix Group: Animal Feed
Legal Information
BCR is a registered trademark of European Commission
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service