Skip to Content
Merck
All Photos(1)

Key Documents

BCR648649

Bovine liver (beta-agonist)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€356.00

€356.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€356.00

About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

€356.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

application(s)

food and beverages

format

matrix material

storage temp.

−20°C

Packaging

set with 2x10 g

Analysis Note

For more information please see:
BCR648649

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Antibiotics: Clenbuterol (free base), Salbutamol, Terbutaline

Matrix type: Meat

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service