Skip to Content
Merck
All Photos(1)

Key Documents

96446

Supelco

N,N′-(Propane-13C3)-1,2-diyldibenzamide-1-15N

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€58.20

€58.20


Estimated to ship on17 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€58.20

About This Item

Empirical Formula (Hill Notation):
13C3C14H1815NNO2
Molecular Weight:
286.31
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€58.20


Estimated to ship on17 April 2025


Request a Bulk Order

grade

analytical standard

Quality Level

isotopic purity

≥99.0%

product line

PESTANAL®

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

−20°C

SMILES string

[13CH3][13CH]([13CH2][15NH]C(C1=CC=CC=C1)=O)NC(C2=CC=CC=C2)=O

InChI

1S/C17H18N2O2/c1-13(19-17(21)15-10-6-3-7-11-15)12-18-16(20)14-8-4-2-5-9-14/h2-11,13H,12H2,1H3,(H,18,20)(H,19,21)/i1+1,12+1,13+1,18+1

InChI key

YHRGPZZLWDLNKT-WHOHSOHCSA-N

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service