Skip to Content
Merck
All Photos(1)

Key Documents

96189

Sigma-Aldrich

Bis(pentafluorophenyl) oxalate

BioReagent, suitable for chemiluminescence

Synonym(s):

Pentafluorophenyl oxalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14F10O4
CAS Number:
Molecular Weight:
422.13
Beilstein:
2068395
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

BioReagent

suitability

suitable for chemiluminescence

SMILES string

Fc1c(F)c(F)c(OC(=O)C(=O)Oc2c(F)c(F)c(F)c(F)c2F)c(F)c1F

Analysis Note

Chemiluminescence: Emission: 430 nm (reaction of TCPO with 9,10-Diphenylanthracene and H2O2)

Other Notes

Reagent for a sensitive detn. method of fluorescent compds. by peroxyoxalate ester chemiluminescence in HPLC and CE

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glenn M Eisen et al.
Gastrointestinal endoscopy, 55(6), 687-694 (2002-04-30)
Chromoendoscopy may reliably separate adenomatous from nonadenomatous polyps. The aim of this multicenter trial was to determine the accuracy of high-resolution chromoendoscopy for the determination of colonic polyp histology. This multicenter trial included 4 academic centers and a primary care
H Yamada et al.
Biomedical chromatography : BMC, 14(5), 333-337 (2000-08-29)
A new sensitive determination method of estradiol in a plasma sample using peroxyoxalate ester chemiluminescence was developed. Estradiol, which was extracted by liquid-liquid extraction using ethyl acetate from plasma, was derivatized with dansyl-chloride (DNS-Cl) and separated by reverse-phase HPLC. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service