Skip to Content
Merck
All Photos(1)

Key Documents

95582

Supelco

Xanthydrol solution

~10% in methanol, for the detection of urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10O2
CAS Number:
Molecular Weight:
198.22
Beilstein:
10395
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

form

liquid

Quality Level

quality

for the detection of urea

concentration

~10% in methanol

density

0.83 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

OC1c2ccccc2Oc3ccccc13

InChI

1S/C13H10O2/c14-13-9-5-1-3-7-11(9)15-12-8-4-2-6-10(12)13/h1-8,13-14H

InChI key

JFRMYMMIJXLMBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kumiko Yamazaki et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 29(5), 705-715 (2012-01-20)
A novel GC-MS method was developed for the determination of acrylamide, which is applicable to a variety of processed foods, including potato snacks, corn snacks, biscuits, instant noodles, coffee, soy sauces and miso (fermented soy bean paste). The method involves
P V Biles et al.
Journal of AOAC International, 81(6), 1155-1161 (1998-12-16)
Contamination of food and food packaging material by rodent urine is evidence of insanitary conditions. Urea from rodent urine is used as a chemical indicator of contamination. The limit of detection of the xanthydrol/urea AOAC Method 959.14 by formation of
Shona Clark et al.
Journal of chromatography. A, 1161(1-2), 207-213 (2007-06-16)
A high-performance liquid chromatography (HPLC) method for the determination of urea that incorporates automated derivatisation with xanthydrol (9H-xanthen-9-ol) is described. Unlike the classic xanthydrol approach for the determination of urea, which involves the precipitation of dixanthylurea (N,N'-di-9H-xanthen-9-ylurea), the derivatisation procedure

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service