Skip to Content
Merck
All Photos(1)

Documents

95135

Supelco

cis-11-Hexadecenal

analytical standard

Synonym(s):

(Z)-11-HDAL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH=CH(CH2)9CHO
CAS Number:
Molecular Weight:
238.41
Beilstein:
1867354
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.3% water

refractive index

n20/D 1.454-1.456
n20/D 1.455 (lit.)

density

0.963 g/mL at 20 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

[H]C(=O)CCCCCCCCC\C=C/CCCC

InChI

1S/C16H30O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h5-6,16H,2-4,7-15H2,1H3/b6-5-

InChI key

AMTITFMUKRZZEE-WAYWQWQTSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

141.8 °F - closed cup

Flash Point(C)

61 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

José G Crespo et al.
Animal behaviour, 98, 79-85 (2014-11-12)
Animal activities, such as foraging and reproduction, are constrained by decisions about how to allocate energy and time efficiently. Overall, male moths invest less in reproduction than females, but they are thought to engage in a scramble competition for access

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service