Skip to Content
Merck
All Photos(1)

Key Documents

92870

Sigma-Aldrich

1,4,10-Trioxa-7,13-diaza-cyclopentadecane

purum, ≥95.0% (GC)

Synonym(s):

Kryptofix® 21

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H22N2O3
CAS Number:
Molecular Weight:
218.29
Beilstein:
607391
EC Number:
MDL number:
UNSPSC Code:
12350000

grade

purum

Assay

≥95.0% (GC)

mp

~80 °C
88-91 °C (lit.)

SMILES string

C1COCCNCCOCCOCCN1

InChI

1S/C10H22N2O3/c1-5-13-6-2-12-4-8-15-10-9-14-7-3-11-1/h11-12H,1-10H2

InChI key

STHIZMRUXPMSCW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Synthesis of disubstituted derivatives[1][2]

Legal Information

Kryptofix is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. Tsukube et al.
The Journal of Organic Chemistry, 56, 268-268 (1991)
I.O. Sutherland
Chemical Society Reviews, 15, 63-63 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service