Skip to Content
Merck
All Photos(1)

Key Documents

92301

Millipore

Listeria mono Selective Supplement I

suitable for microbiology

Synonym(s):

Polymyxin B sulfate salt, Listeria monocytogenes Selective Supplement I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C55H96N16O13 · 2H2SO4
CAS Number:
Molecular Weight:
1385.61
EC Number:
MDL number:
UNSPSC Code:
41106200
NACRES:
NA.85

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
fungi

application(s)

environmental
food and beverages

microbiology

Mode of action

cell membrane | interferes

storage temp.

2-8°C

suitability

Listeria spp.

SMILES string

O=C(C(NC(C(CCN)NC(CCCCC(C)CC)=O)=O)C(C)O)NC(CCN)C(NC(CCNC(C(NC(C(NC(C(CCN)N1)=O)CCN)=O)C(O)C)=O)C(NC(CCN)C(NC(CC2=CC=CC=C2)C(NC(CC(C)C)C1=O)=O)=O)=O)=O.O=S(O)(O)=O

InChI

1S/C48H82N16O13.H2O4S/c1-27(2)24-37-47(76)59-32(11-19-52)41(70)56-31(10-18-51)43(72)61-35(14-22-65)39(68)54-21-13-34(45(74)57-33(12-20-53)44(73)64-38(48(77)63-37)25-28-6-4-3-5-7-28)60-42(71)30(9-17-50)58-46(75)36(15-23-66)62-40(69)29(8-16-49)55-26-67;1-5(2,3)4/h3-7,26-27,29-38,65-66H,8-25,49-53H2,1-2H3,(H,54,68)(H,55,67)(H,56,70)(H,57,74)(H,58,75)(H,59,76)(H,60,71)(H,61,72)(H,62,69)(H,63,77)(H,64,73);(H2,1,2,3,4)

InChI key

HNDFYNOVSOOGDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A selective supplement recommended by ISO Committee for the isolation of Listeria species.
Chemical structure: peptide

Application

for the isolation of Listeria species

Biochem/physiol Actions

Mode of Action: Polymyxin B Sulfate binds to the lipid A portion of bacterial lipopolysaccharides†, disrupting the cytoplasmic membrane by inducing pores large enough to permit nucleotide leakage in bacterial walls. This disrupts the permeability of the cytoplasmic membrane.

Antimicrobial spectrum: Has bactericidal action on most gram-negative bacilli††, including E. Coli and on most fungi and gram-positive bacteria.

Components

(per vial, sufficient for 500 ml medium)
Polymyxin B sulphate 38350 IU

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kensuke Nakamura et al.
The American journal of emergency medicine, 31(5), 893-893 (2013-02-13)
Septic patients often have low cardiac output. Some of them present severe cardiac dysfunction such as septic cardiomyopathy. However, no well-known and effective treatment for septic cardiomyopathy exists. The effect of endotoxin adsorption by polymyxin B–immobilized fiber column–direct hemoperfusion (PMX-DHP)
Conjugated polymer-coated bacteria for multimodal intracellular and extracellular anticancer activity.
Chunlei Zhu et al.
Advanced materials (Deerfield Beach, Fla.), 25(8), 1203-1208 (2013-01-03)
Lee Sun New et al.
Analytical and bioanalytical chemistry, 405(4), 1407-1415 (2012-11-28)
With increasing multidrug resistance coupled to a poor development pipeline, clinicians are exploring antimicrobial combinations to improve treatment outcomes. In vitro hollow-fiber infection model (HFIM) is employed to simulate human in vivo drug clearance and investigate pharmacodynamic synergism of antibiotics.
The novel polymyxin derivative NAB739 is remarkably less cytotoxic than polymyxin B and colistin to human kidney proximal tubular cells.
Martti Vaara et al.
International journal of antimicrobial agents, 41(3), 292-293 (2012-11-28)
Naoki Sawa et al.
Clinical nephrology, 79(6), 463-470 (2012-12-21)
Vasopressin and direct hemoperfusion with an immobilized polymyxin B column (PMX) have emerged recently as treatments for septic shock. This study assessed the impact of these two treatments on the survival of patients with septic shock. A retrospective, matched cohort

Protocols

Listeria is a public health concern, affecting elderly, pregnant, newborns, and immunocompromised individuals.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service