Skip to Content
Merck
All Photos(2)

Documents

916625

Sigma-Aldrich

tert-Butyl 4-(3-(2-aminoethyl)phenyl)piperidine-1-carboxylate

≥95%

Synonym(s):

Semi-flexible linker for PROTAC® development

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H28N2O2
CAS Number:
Molecular Weight:
304.43
UNSPSC Code:
12352116

Quality Level

Assay

≥95%

form

liquid

reaction suitability

reagent type: linker

functional group

Boc
amine

storage temp.

2-8°C

SMILES string

O=C(N(CC1)CCC1C2=CC(CCN)=CC=C2)OC(C)(C)C

Application

tert-Butyl 4-(3-(2-aminoethyl)phenyl)piperidine-1-carboxylate is a 4-aryl piperidine useful as a semi-flexible linker in PROTAC development for targeted protein degradation. Incorporation of rigidity into the linker region of bifunctional protein degraders may impact the 3D orientation of the degrader and thus ternary complex formation as well as optimization of drug-like properties.

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service