Skip to Content
Merck
All Photos(1)

Documents

91274

Millipore

Penicillinase Concentrate

suitable for (Antibiotic inactivation), vial of 100 mL

Synonym(s):

Penicillinase solution, β-Lactamase I, β-Lactamase I concentrate, Penicillinase enzyme

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41171614
NACRES:
NA.76

form

liquid

Quality Level

shelf life

limited shelf life, expiry date on the label

packaging

vial of 100 mL

suitability

suitable for (Antibiotic inactivation)

application(s)

microbiology

storage temp.

2-8°C

General description

Penicillinase Concentrate is a highly purified enzyme preparation that contains β-Lactamase, which is also known as penicillinase. It is a ready-to-use solution that can be directly added to test samples (for the inactivation of beta-lactams from blood or tissue samples before routine microbiological examination) or can be used in media preparation (for either environmental monitoring or antibiotic sterility testing). This concentrate is specifically designed for use in microbiology as a supplement to culture media to deactivate antibiotics in microbial samples. It can inactivate Penicillin G, one of the commonly used Penicillin, at an impressive rate of 10,000,000 International Units (IU) per mL per minute.

Penicillinase Concentrate is a valuable tool for hydrolyzing and inactivating the β-lactam ring found in penicillin and other related antibiotics. It plays a critical role in studying antibiotic resistance mechanisms and in the development of antibiotic detection assays.

Application

PenicillinaseConcentrate is a culture media supplement used for the deactivation of antibioticsin microbial samples.


Inactivation of Penicillin G* 10,000,000 IU/mL/min


*Not all penicillins are comparable to Penicillin G in regards to their inactivation by penicillinase. Some may require as much as 20 times more penicillinase than does Penicillin G


Potency** 20,000 Levy units/ml/min


**Levy Unit (LU): amount of penicillinase that inactivates 59.3 IU( International Units) of Sodium Penicillin G per minute at 25°C and pH 7.0

Packaging

100 ml per vial

Components

Composition:
(100 ml per vial)

Inactivation of Penicillin G* 10,000,000 IU/mL/min
*Not all penicillins are comparable to Penicillin G in regards to their inactivation by penicillinase. Some may require as much as 20 times more penicillinase than does Penicillin G.

Potency** 20,000 Levy units/ml/min
**Levy Unit (LU): amount of penicillinase that inactivates 59.3 IU( International Units) of Sodium Penicillin G per minute at 25°C and pH 7.0

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service