Skip to Content
Merck
All Photos(1)

Documents

87390

Sigma-Aldrich

2(3)-(Tetrahydrofurfuryloxy)tetrahydropyran

technical, ≥90% (GC)

Synonym(s):

‘T-Solvent’

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O3
CAS Number:
Molecular Weight:
186.25
Beilstein:
109779
EC Number:
MDL number:
UNSPSC Code:
12352100

grade

technical

Assay

≥90% (GC)

refractive index

n20/D 1.461 (lit.)
n20/D 1.465

bp

235-238 °C

density

1.03 g/mL at 25 °C (lit.)

SMILES string

C1CCC(OC1)OCC2CCCO2.C3COCC(C3)OCC4CCCO4

InChI

1S/2C10H18O3/c1-3-9(7-11-5-1)13-8-10-4-2-6-12-10;1-2-6-12-10(5-1)13-8-9-4-3-7-11-9/h2*9-10H,1-8H2

InChI key

OMNURHQDSBSXKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

solvent for LiAH4-reduction

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service