Skip to Content
Merck
All Photos(2)

Key Documents

81610

Supelco

Poly(styrenesulfonic acid sodium salt)

analytical standard, for GPC, 32,000

Synonym(s):

Poly(sodium styrenesulfonate)

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€101.00

€101.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€101.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€101.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard
for GPC

Quality Level

shelf life

limited shelf life, expiry date on the label

mol wt

Mp ~30900
Mw ~32000

technique(s)

gel permeation chromatography (GPC): suitable

Mw/Mn

~1.1

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Poly(styrenesulfonic acid sodium salt) is used as an analytical size standard in the following:
  • Comparing poly(styrenesulfonate) (NaPSS) chain ordering between two solid interfaces and in bulk using colloidal probe atomic force microscopy (CP-AFM) and small-angle X-ray scattering (SAXS).[1]
  • The synthesis of biohybrid materials of a stimulus-responsive hydrogel based on such a ligand-responsive DNA-protein interaction.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Conditional DNA-Protein Interactions Confer Stimulus-Sensing Properties to Biohybrid Materials
Christen, E., H., et al.
Advances in Functional Materials, 21(15), 2861-2867 (2011)
Structuring of polyelectrolyte (NaPSS) solutions in bulk and under confinement as a function of concentration and molecular weight
Uzum C, et al.
Macromolecules, 44(19), 7782-7791 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service