Skip to Content
Merck
All Photos(1)

Key Documents

81416

Supelco

Polystyrene

analytical standard, for GPC, 1,000,000

Synonym(s):

PS

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€109.00

€109.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
250 MG
€109.00

About This Item

Linear Formula:
[CH2CH(C6H5)]n
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€109.00


Available to ship on31 March 2025Details


Request a Bulk Order

grade

analytical standard
for GPC

Quality Level

shelf life

limited shelf life, expiry date on the label

mol wt

Mn ~930000
Mp ~1000000
Mw ~960000

technique(s)

gel permeation chromatography (GPC): suitable

Mw/Mn

~1.03

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

  1. What is polydispersity of this polystyrene?

    1 answer
    1. The polydispersity is not determined.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service