79895
(2RS)-2-(4-Formylphenyl)propionic acid
pharmaceutical impurity standard
Synonym(s):
(2RS)-2-(4-Formylphenyl)propanoic acid
About This Item
Recommended Products
Quality Level
Assay
≥98.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
impurities
≤1.0% water
application(s)
clinical testing
format
neat
SMILES string
OC(C(C)C1=CC=C(C([H])=O)C=C1)=O
InChI
1S/C10H10O3/c1-7(10(12)13)9-4-2-8(6-11)3-5-9/h2-7H,1H3,(H,12,13)
InChI key
IAXYHYOWEQQFMC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Application in enzyme inhibition studies: (2RS)-2-(4-Formylphenyl)propionic acid is utilized to explore enzyme interactions and inhibitory mechanisms within pharmaceutical and biochemical research, providing insights that assist in the development of therapeutic agents and drugs (Ploch-Jankowska, 2024).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service