Skip to Content
Merck
All Photos(1)

Key Documents

75607

Sigma-Aldrich

Trimethyl orthovalerate

purum, ≥97.0% (GC)

Synonym(s):

1,1,1-Trimethoxypentane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH2C(OCH3)3
CAS Number:
Molecular Weight:
162.23
Beilstein:
1739200
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.410 (lit.)
n20/D 1.410

bp

164-166 °C (lit.)

density

0.941 g/mL at 25 °C (lit.)

SMILES string

CCCCC(OC)(OC)OC

InChI

1S/C8H18O3/c1-5-6-7-8(9-2,10-3)11-4/h5-7H2,1-4H3

InChI key

XUXVVQKJULMMKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trimethyl orthovalerate is used in the synthesis of 1,2,4-triazolo[1,5-c]pyrimidine derivatives as human adenosine A3 receptor ligands.[1] It can also be used as a precursor to prepare N-sulfonyl and N-sulfinyl imines and imidates.[2]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Facile synthesis of fused 1, 2, 4-triazolo [1, 5-c] pyrimidine derivatives as human adenosine A 3 receptor ligands.
Okamura T, et al.
Bioorganic & Medicinal Chemistry Letters, 14(10), 2443-2446 (2004)
Metal Free, Microwave Assisted Preparation of N-Sulfonyl and N-Sulfinyl Imines and Imidates.
Verrier C, et al.
ACS sustainable chemistry & engineering, 14(10), 2443-2446 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service