Skip to Content
Merck
All Photos(1)

Key Documents

70049

Sigma-Aldrich

Myrcene

technical, ~90% (GC)

Synonym(s):

β-Myrcene, 7-Methyl-3-methylene-1,6-octadiene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHC(=CH2)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
136.23
Beilstein:
1719990
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

4.7 (vs air)

vapor pressure

~7 mmHg ( 20 °C)

grade

technical

Assay

~90% (GC)

refractive index

n20/D 1.469 (lit.)
n20/D 1.469

bp

167 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

SMILES string

C\C(C)=C/CCC(=C)C=C

InChI

1S/C10H16/c1-5-10(4)8-6-7-9(2)3/h5,7H,1,4,6,8H2,2-3H3

InChI key

UAHWPYUMFXYFJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Rivoal et al.
Chemosphere, 78(8), 942-949 (2010-01-23)
The large amount of volatile organic compound (VOC) emitted by vegetation modifies air quality contributing to both tropospheric ozone and secondary organic aerosol production. A better understanding of the factors controlling VOC emissions by vegetation is mandatory in order to
Flavia Bonamin et al.
Chemico-biological interactions, 212, 11-19 (2014-02-01)
The monoterpene β-myrcene has been widely used in cosmetics, food and beverages, and it is normally found in essential oil from citrus fruit. The aim of this study was to investigate the anti-ulcer effects of β-myrcene on experimental models of
Mark L Thompson et al.
Applied microbiology and biotechnology, 85(3), 721-730 (2009-08-27)
The biocatalytic generation of high-value chemicals from abundant, cheap and renewable feedstocks is an area of great contemporary interest. A strain of Rhodococcus erythropolis designated MLT1 was isolated by selective enrichment from the soil surrounding hop plants, using the abundant
Akbar Esmaeili et al.
Chemistry Central journal, 5, 26-26 (2011-05-26)
Dihydrolinalool and terpineol are sources of fragrances that provide a unique volatile terpenoid alcohol of low toxicity and thus are widely used in the perfumery industry, in folk medicine, and in aromatherapy. They are important chemical constituents of the essential
Johannes Stökl et al.
Proceedings. Biological sciences, 278(1709), 1216-1222 (2010-10-15)
Most insects are dependent on chemical communication for activities such as mate finding or host location. Several plants, and especially orchids, mimic insect semiochemicals to attract insects for unrewarded pollination. Here, we present a new case of pheromone mimicry found

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service