Skip to Content
Merck
All Photos(1)

Documents

69184

Supelco

4-Phenylphenol

reference material

Synonym(s):

4-Hydroxybiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4OH
CAS Number:
Molecular Weight:
170.21
Beilstein:
1907452
MDL number:
UNSPSC Code:
41116107

grade

reference material

Quality Level

Assay

≥96.0% (GC)

shelf life

limited shelf life, expiry date on the label

bp

321 °C (lit.)

mp

164-166 °C (lit.)

format

neat

SMILES string

Oc1ccc(cc1)-c2ccccc2

InChI

1S/C12H10O/c13-12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9,13H

InChI key

YXVFYQXJAXKLAK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service