Skip to Content
Merck
All Photos(1)

Key Documents

67575

Supelco

4-Chlorobenzyl chloride

analytical standard

Synonym(s):

α,4-Dichlorotoluene, (4-Chlorophenyl)methyl chloride, 1-Chloro-4-(chloromethyl)benzene, NSC 34032

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH2Cl
CAS Number:
Molecular Weight:
161.03
Beilstein:
471558
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

216-222 °C (lit.)

mp

27-29 °C (lit.)

application(s)

environmental
pharmaceutical

format

neat

SMILES string

ClCc1ccc(Cl)cc1

InChI

1S/C7H6Cl2/c8-5-6-1-3-7(9)4-2-6/h1-4H,5H2

InChI key

JQZAEUFPPSRDOP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Chlorobenzyl chloride is generally used in the preparation of quaternary ammonium salts. It acts as an intermediate in the production of rice herbicide, pharmaceuticals and pesticides.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A catalytic method for the selective chlorination of benzyl chloride to 4-chlorobenzyl chloride using zeolite catalysts
Kale SM and Singh AP
J. Mol. Catal. A: Chem., 138(2-3), 263-272 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service