Skip to Content
Merck
All Photos(1)

Documents

64397

Supelco

α-Amylcinnamaldehyde

mixture of cis and trans, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=C[(CH2)4CH3]CHO
CAS Number:
Molecular Weight:
202.29
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.557 (lit.)
n20/D 1.557

bp

287-290 °C (lit.)

density

0.97 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

[H]C(/C(CCCCC)=C/C1=CC=CC=C1)=O

InChI

1S/C14H18O/c1-2-3-5-10-14(12-15)11-13-8-6-4-7-9-13/h4,6-9,11-12H,2-3,5,10H2,1H3/b14-11+

InChI key

HMKKIXGYKWDQSV-SDNWHVSQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

α-Amylcinnamaldehyde may be used as a reference standard for the determination of α-amylcinnamaldehyde in cosmetic creams by direct contact sorptive tape extraction–gas chromatography–mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

294.8 °F

Flash Point(C)

146 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative determination of some volatile suspected allergens in cosmetic creams spread on skin by direct contact sorptive tape extraction?gas chromatography?mass spectrometry.
Sgorbini B, et al.
Journal of Chromatography A, 1217(16), 2599-2605 (2010)
Eiram N Elahi et al.
Chemical research in toxicology, 17(3), 301-310 (2004-03-18)
Skin protein modification (haptenation) is thought to be a key step in the manifestation of sensitization to low molecular mass chemicals (<500 g/mol). For sensitizing chemicals that are not protein reactive, it is hypothesised that metabolic activation can convert such
J D Guin et al.
Journal of the American Academy of Dermatology, 8(1), 76-80 (1983-01-01)
Sensitivity to alpha-amylcinnamic aldehyde (alpha-AcAld) is apparently uncommon, but, like allergy to alpha-amylcinnamic alcohol (alpha-AcAlc), it often accompanies allergy to the perfume in Mycolog cream. Although alpha-AcAlc is a known ingredient, alpha-AcAld is not. However, gas-liquid chromatographic analysis shows alpha-AcAld
S C Rastogi et al.
Contact dermatitis, 38(1), 29-35 (1998-03-21)
Deodorants are one of the most frequently used types of cosmetics and side-effects from them are common. Recent studies relate perfume allergy to this type of product. 73 deodorants were analyzed by gas chromatography--mass spectrometry for the determination of the
S C Rastogi et al.
Contact dermatitis, 34(6), 423-426 (1996-06-01)
In the present study, we have investigated 42 cosmetic products based on natural ingredients for content of 11 fragrance substances: geraniol, hydroxycitronellal, eugenol, isoeugenol, cinnamic aldehyde, cinnamic alcohol, alpha-amylcinnamic aldehyde, citral, coumarin, dihydrocoumarin and alpha-hexylcinnamic aldehyde. The study revealed that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service