Skip to Content
Merck
All Photos(1)

Key Documents

603295

Supelco

Caffeine-13C3 solution

99 atom % 13C, 1 mg/mL in methanol, drug standard

Synonym(s):

Caffeine-trimethyl-13C3 solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3C5H10N4O2
Molecular Weight:
197.17
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

drug standard

Quality Level

isotopic purity

99 atom % 13C

availability

not available in Japan

concentration

1 mg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

[13CH3]N1C(=O)N([13CH3])c2ncn([13CH3])c2C1=O

InChI

1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3/i1+1,2+1,3+1

InChI key

RYYVLZVUVIJVGH-VMIGTVKRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Caffeine-13C3 solution can be used as an internal standard for the analysis of contaminants of emerging concern (CECs)in the influent and effluent streams of wastewater treatment plants using liquid chromatographic technique coupled with high resolution mass spectrometry(LC-HRMS) and liquid chromatographic technique coupled with tandem mass spectrometry(LC-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Estimating removals of contaminants of emerging concern from wastewater treatment plants: The critical role of wastewater hydrodynamics
Baalbaki Z, et al.
Chemosphere, 178, 439-448 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service