Skip to Content
Merck
All Photos(1)

Key Documents

570753

Sigma-Aldrich

Dibenzo[f,h]quinoline

Synonym(s):

1-Aza-triphenylene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H11N
CAS Number:
Molecular Weight:
229.28
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

solid

mp

184.0-185.9 °C (lit.)

λmax

257 nm in chloroform

application(s)

hematology
histology

storage temp.

room temp

SMILES string

c1ccc2c(c1)c3ccccc3c4ncccc24

InChI

1S/C17H11N/c1-2-7-14-12(6-1)13-8-3-4-9-15(13)17-16(14)10-5-11-18-17/h1-11H

InChI key

RIYPENPUNLHEBK-UHFFFAOYSA-N

Application

Used in the preparation of near infrared luminescent metal (Yb3+, Nd3+, Er3+) azatriphenylene complexes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service