Skip to Content
Merck
All Photos(1)

Key Documents

55578

Supelco

4-Hydroxyisovaleric acid sodium salt hydrate

analytical standard

Synonym(s):

3-Methyl-γ-hydroxybutyric acid sodium salt hydrate, 4-Hydroxy-3-methylbutanoic acid sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O3 · xNa+ · yH2O
CAS Number:
Molecular Weight:
118.13 (anhydrous free acid basis)
MDL number:
UNSPSC Code:
12352200

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)
80.0-105.0% (free acid, anhydrous, NT)

shelf life

limited shelf life, expiry date on the label

impurities

≤25% water

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

OC(CC(C)CO)=O

InChI

1S/C5H10O3/c1-4(3-6)2-5(7)8/h4,6H,2-3H2,1H3,(H,7,8)

InChI key

ZGYGEPZZMAXSKH-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service