Skip to Content
Merck
All Photos(1)

Key Documents

55441

Supelco

Ethyl oleate

analytical standard

Synonym(s):

Oleic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COOC2H5
CAS Number:
Molecular Weight:
310.51
Beilstein:
1727318
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.451 (lit.)

bp

216-218 °C/15 mmHg

mp

−32 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCC(=O)OCC

InChI

1S/C20H38O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h11-12H,3-10,13-19H2,1-2H3/b12-11-

InChI key

LVGKNOAMLMIIKO-QXMHVHEDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rosa Palagano et al.
Foods (Basel, Switzerland), 9(7) (2020-07-18)
The content of fatty acid ethyl esters (FAEEs) is one of the quality parameters to define if an olive oil can be classified as extra virgin as these compounds are considered markers for virgin olive oils obtained from poor-quality olives.
Elisabetta Bertol et al.
Drug testing and analysis, 6(9), 930-935 (2014-01-17)
Fatty acid ethyl esters (FAEEs) are minor ethanol metabolites that can accumulate in hair. The performance of hair FAEEs as a biomarker that can discriminate null or moderate drinking from risky, excessive drinking was verified by evaluating the relationship between
Jiangbo Hu et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 88(2), 556-564 (2014-06-26)
The purpose of this study was to develop and evaluate the supermolecular evodiamine (EVO) loaded water-in-oil nanoemulsions containing brucea javanica oil (NESEB) with enhanced physicochemical and biological characteristics. NESEB was fabricated by applying supermolecular phytosome nanotechnology and nanoemulsification technology together
Simona Pichini et al.
Clinical chemistry and laboratory medicine, 52(8), 1179-1185 (2014-03-13)
Ethyl glucuronide (EtG) measurement in neonatal meconium has emerged as a reliable marker to objectively assess prenatal exposure to maternal ethanol complementary to fatty acid ethyl ester (FAEEs) measurement. The detection of EtG in meconium is currently a lengthy, difficult
Jianbin Zhang et al.
AAPS PharmSciTech, 15(3), 731-740 (2014-02-21)
The objective of this study is to investigate the effect of lipolysis on the release of poorly water-soluble drug from SMEDDS in the perspective of drug core/shell location. For this purpose, four SMEDDS formulations with various core/shell properties were developed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service