Skip to Content
Merck
All Photos(1)

Documents

53940

Supelco

Hydrindantin dihydrate

for Stein-Moore-Chromatography, LiChropur

Synonym(s):

2,2′,3,3,3′,3′-Hexahydroxy-2,2′-biindan-1,1′-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14O8
CAS Number:
Molecular Weight:
358.30
Beilstein:
1895666
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for Stein-Moore-Chromatography

Quality Level

quality

LiChropur

technique(s)

HPLC: suitable

mp

~240 °C (dec.)

SMILES string

[H]O[H].[H]O[H].OC1(O)c2ccccc2C(=O)C1(O)C3(O)C(=O)c4ccccc4C3(O)O

InChI

1S/C18H10O6.2H2O/c19-13-9-5-1-2-6-10(9)14(20)17(13,23)18(24)15(21)11-7-3-4-8-12(11)16(18)22;;/h1-8,23-24H;2*1H2

InChI key

QIAIXVRAKYAOGJ-UHFFFAOYSA-N

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service