Skip to Content
Merck
All Photos(1)

Key Documents

53183

Supelco

Metformin-(dimethyl-d6) hydrochloride

analytical standard

Synonym(s):

1,1-Di(methyl-d3)biguanide hydrochloride, [2H6]-Metformin hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€453.00

€453.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
5 MG
€453.00

About This Item

Empirical Formula (Hill Notation):
C4D6H5N5 · HCl
CAS Number:
Molecular Weight:
171.66
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€453.00


In StockDetails


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

storage temp.

2-8°C

SMILES string

NC(NC(N(C([2H])([2H])[2H])C([2H])([2H])[2H])=N)=N.Cl

InChI

1S/C4H11N5.ClH/c1-9(2)4(7)8-3(5)6;/h1-2H3,(H5,5,6,7,8);1H/i1D3,2D3;

InChI key

OETHQSJEHLVLGH-TXHXQZCNSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

Identification (LC-MS): identity conforms

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kiran Chaudhari et al.
PloS one, 15(6), e0234571-e0234571 (2020-06-12)
Metformin, an anti-diabetes drug, has been recently emerging as a potential "anti-aging" intervention based on its reported beneficial actions against aging in preclinical studies. Nonetheless, very few metformin studies using mice have determined metformin concentrations and many effects of metformin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service