49757
Tetrahydrofurfuryl alcohol
analytical standard
Synonym(s):
NSC 15434, Tetrahydro-2-furanmethanol
About This Item
Recommended Products
grade
analytical standard
Quality Level
vapor density
3.52 (vs air)
vapor pressure
2.3 mmHg ( 39 °C)
Assay
≥98.5% (GC)
shelf life
limited shelf life, expiry date on the label
expl. lim.
9.7 %
refractive index
n20/D 1.452 (lit.)
n20/D 1.452-1.454
bp
178 °C (lit.)
mp
−80 °C (lit.)
density
1.054 g/mL at 25 °C (lit.)
application(s)
cleaning products
cosmetics
food and beverages
personal care
format
neat
SMILES string
OCC1CCCO1
InChI
1S/C5H10O2/c6-4-5-2-1-3-7-5/h5-6H,1-4H2
InChI key
BSYVTEYKTMYBMK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Repr. 1B
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 2
Flash Point(F)
163.4 °F - closed cup
Flash Point(C)
73 °C - closed cup
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
EU REACH Annex XVII (Restriction List)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service