Skip to Content
Merck
All Photos(3)

Documents

48914-U

Sigma-Aldrich

ProClin 300

pkg of 400 mL, ProClin preservative for use in IVD manufacturing, CMIT/MIT 3 %

Synonym(s):

3% solution of 5-chloro-2-methyl-4-isothiazolin-3-one and 2-Methyl-4-isothiazolin-3-one

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201
NACRES:
NA.21

form

liquid

shelf life

3 yr

contains

Alkyl Carboxylate as stabilizer

composition

CMIT/MIT, 3%

packaging

pkg of 400 mL

parameter

<45 °C operating temperature

color

colorless to light yellow

matrix

Modified glycol

operating pH range

2.5-8.5

pH

4.1 (100 g/L)

bp

189 °C

mp

-40 °C

solubility

water: soluble

density

1.03 g/cm3

Looking for similar products? Visit Product Comparison Guide

Application

ProClin preservatives are immediately microstatic upon contact with microbial organisms as a result of their ability to quickly penetrate cell membranes and inhibit specific enzymes crucial to cellular respiration. Multiple specific sites of ProClin toxicity prevent microorganisms from developing high levels of resistance, making ProClin highly efficient even over extended storage periods of your IVD components.

Legal Information

ProClin is a trademark of LANXESS Corp.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada′s Pest Management Regulatory Agency, Turkey′s Biocidal Products Regulation, Korea′s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

244.4 °F - closed cup

Flash Point(C)

118 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Sats et al.
Journal of dairy science, 103(8), 6810-6819 (2020-06-15)
Bovine colostrum, as vital as it is for calves, is also a valuable source of functional components with rich health benefits for humans. Bovine colostrum whey consists of a large number of bioactive proteins and peptides. The most abundant of
James P Higham et al.
PloS one, 6(7), e22398-e22398 (2011-07-30)
Nutritional status is a critical element of many aspects of animal ecology, but has proven difficult to measure non-invasively in studies of free-ranging animals. Urinary C-peptide of insulin (UCP), a small polypeptide cleaved in an equimolar ratio from proinsulin when
Daesub Song et al.
Journal of clinical microbiology, 53(4), 1178-1182 (2015-01-30)
We present here a rapid immunochromatographic assay for the detection of Middle East respiratory syndrome coronavirus (MERS-CoV) antigen in the nasal swabs of dromedary camels. The assay is based on the detection of MERS-CoV nucleocapsid protein in a short time

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service