Skip to Content
Merck
All Photos(1)

Key Documents

46145

Sigma-Aldrich

Europium(III)-tris-(2,2,6,6-tetramethyl-3,5-heptanedionate)

purum p.a., for NMR spectroscopy

Synonym(s):

Eu(dpm)3, Europium-tris(dipivaloylmethane), Resolve-Al, Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)europium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Eu(OCC(CH3)3CHCOC(CH3)3)3
CAS Number:
Molecular Weight:
701.77
Beilstein:
4167338
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

grade

for NMR spectroscopy
purum p.a.

reaction suitability

core: europium
reagent type: catalyst

mp

187-191 °C

suitability

suitable for NMR (spectroscopy)

storage temp.

2-8°C

SMILES string

CC(C)(C)C(=O)\C=C(/O[Eu](O\C(=C/C(=O)C(C)(C)C)C(C)(C)C)O\C(=C/C(=O)C(C)(C)C)C(C)(C)C)C(C)(C)C

InChI

1S/3C11H20O2.Eu/c3*1-10(2,3)8(12)7-9(13)11(4,5)6;/h3*7,12H,1-6H3;/q;;;+3/p-3/b3*8-7-;

InChI key

DVUACRCEVJAWMS-LWTKGLMZSA-K

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michiya Fujiki et al.
Frontiers in chemistry, 8, 685-685 (2020-09-10)
We report emerging circularly polarized luminescence (CPL) at 4f-4f transitions when lanthanide (EuIII and TbIII) tris(β-diketonate) embedded to cellulose triacetate (CTA), cellulose acetate butyrate (CABu), D-/L-glucose pentamethyl esters ( D-/ L-Glu), and D-/L-arabinose tetramethyl esters ( D-/ L-Ara) are in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service