Skip to Content
Merck
All Photos(2)

Key Documents

45984

Supelco

2-Methyl-5-nitroaniline

analytical standard

Synonym(s):

2-Amino-4-nitrotoluene, 5-Nitro-o-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)NH2
CAS Number:
Molecular Weight:
152.15
Beilstein:
879021
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

103-106 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Cc1ccc(cc1N)[N+]([O-])=O

InChI

1S/C7H8N2O2/c1-5-2-3-6(9(10)11)4-7(5)8/h2-4H,8H2,1H3

InChI key

DSBIJCMXAIKKKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methyl-5-nitroaniline is one of the laser-induced decomposition products of Pigment Red 22 and Pigment Red 9 (widely used azo compound).

Application

2-Methyl-5-nitroaniline may be used as an analytical reference standard for the quantification of the analyte in groundwater samples using gas chromatography combined with electron-capture negative-ion chemical ionization mass spectrometry (GC-EC-NICI-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of aromatic amines at trace levels by derivatization with heptafluorobutyric anhydride and gas chromatography-electron-capture negative-ion chemical ionization mass spectrometry
Longo M and Cavallaro A
Journal of Chromatography A, 753(1), 91-100 (1996)
Determination of aromatic amines at trace levels by derivatization with heptafluorobutyric anhydride and gas chromatography-electron-capture negative-ion chemical ionization mass spectrometry
Engel E, et al.
Analytical Chemistry, 78, 6440-6447 (2006)
5-Nitro-ortho-toluidine.
IARC monographs on the evaluation of carcinogenic risks to humans, 48, 169-177 (1990-01-01)
H Shimizu et al.
Gut, 50(2), 266-270 (2002-01-15)
5-Nitro-o-toluidine is an aromatic nitro amino compound. While other aromatic compounds are known to damage the human liver and are registered as toxic substances, toxicity information concerning 5-nitro-o-toluidine is lacking. To investigate the hepatotoxicity of 5-nitro-o-toluidine. Of 15 workers in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service